Anti-DYRK3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-9520R-CY7
10490-698EA 792.04 USD
10490-698
Anti-DYRK3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Dyrk (for dual specificity tyrosine phosphorylation regulated kinase) is the homolog of the Drosophila mnb (minibrain) gene, which is required for neurogenesis. Dyrk is a dual-specificity tyrosine kinase and serine/threonine kinase, which is itself regulated by tyrosine phosphorylation. Several mammalian Dyrk related proteins have been identified and are thought to compose a family of dual specificity protein kinases. Dyrk family members, including Dyrk1A (originally Dyrk), Dyrk1B, Dyrk1C, Dyrk2, Dyrk3, Dyrk4A and Dyrk4B, are thought to be involved in diverse cellular functions. Two isoforms of human fetal brain Dyrk2 exist: a deduced 528-amino acid protein and a protein containing 73 additional amino acids at the amino terminus. Dyrk3 is strongly expressed in testis, only after the onset of spermatogenesis, and very weakly expressed in spleen and adrenal gland. The genes which encode Dyrk2 and Dyrk3 map to human chromosomes 12 and 1q32, respectively.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DYRK3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR