Anti-BPGM Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-9519R-A647
10490-664EA 792.04 USD
10490-664
Anti-BPGM Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
BPGM (2,3-bisphosphoglycerate mutase) is a 259 amino acid protein that belongs to the phosphoglycerate mutase family and exists as a homodimer that plays a crucial role in the regulation of hemoglobin oxygen. Specifically, BPGM catalyzes the conversion of 3-D-glyceroyl phosphate to 2,3-bisD-glycerate (2,3-BPG), a reaction that is essential for controlling the concentration of 2,3-BPG within the cell. The gene encoding BPGM maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. Involvement in disease:Defects in BPGM are the cause of bisphosphoglycerate mutase deficiency (BPGMD) . A disease characterized by hemolytic anemia, splenomegaly, cholelithiasis and cholecystitis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BPGM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR