Anti-AHSG Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-9518R-A555
10490-654EA 792.04 USD
10490-654
Anti-AHSG Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Act as a chaperone to prevent the harmful aggregation of alpha-hemoglobin during normal erythroid cell development. Specifically protects free alpha-hemoglobin from precipitation. It is predicted to modulate pathological states of alpha-hemoglobin excess such as beta-thalassemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AHSG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR