Anti-RGAG4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-8501R-A350
10485-910EA 792.04 USD
10485-910
Anti-RGAG4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
RGAG4

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGAG4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR