Anti-RFTN2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-8500R-A555
10485-898EA 792.04 USD
10485-898
Anti-RFTN2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Membrane microdomains known as lipid rafts are implicated in B-cell activation during B-cell receptor (BCR) signal initiation. Raftlin-2, also designated RFTN2 (raftlin family member 2) or raft-linking protein 2, is a 501 amino acid cell membrane protein that is essential for raft cell assembly and maintenance. A lipid anchor protein, Raftlin-2 belongs to the raftlin family and is encoded by a gene that maps to human chromosome 2q33.1 and mouse chromosome 1 C1.2. Human chromosome 2 is the second largest human chromosome, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8. An extremely rare recessive genetic disorder, Alstré°‰ syndrome is due to mutations in the ALMS1 gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RFTN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR