Anti-DYRK3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-8499R-CY7
10485-886EA 791.8 USD
10485-886
Anti-DYRK3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The function of RED is currently unknown. The protein encoded by the RED gene was identified by its RED repeat, a stretch of repeated arginine, glutamic acid and aspartic acid residues. The protein localizes to discrete dots within the nucleus, excluding the nucleolus. This gene maps to chromosome 5; however, a pseudogene may exist on chromosome 2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DYRK3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR