Anti-RENBP Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-8497R-CY3
10485-832EA 792.04 USD
10485-832
Anti-RENBP Rabbit Polyclonal Antibody (Cy3®)
Antibodies
RENBP is a 427 amino acid protein that is a proteinaceous renin inhibitor. In inhibiting renin, RENBP forms a complex with it; a high molecular weight renin. RENBP contains a leucine zipper domain, which is essential for its dimerization with renin. RENBP can catalyze the interconversion of N-acetylglucosamine to N-acetylmannosamine, indicating that it is a GlcNAc 2-epimerase. Sequences of porcine, human and rat renin-binding proteins are highly homologous. The RENBP gene is conserved in dog, mouse, rat and zebrafish, and maps to human chromosome Xq28 between DXS52 and G6PD. Rat Renbp is located on chromosome X at Xq37 close to marker DXWox3 and falls outside the BP QTL regions on chromosome X.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RENBP
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR