Anti-RNF210 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-8494R-CY5.5
10485-764EA 792.04 USD
10485-764
Anti-RNF210 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
RNF210

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF210
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR