Anti-FBXO25 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-8491R-FITC
10485-512EA 776.97 USD
10485-512
Anti-FBXO25 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
FBXO25 is a member of the F box protein family which is characterized by an approximately 40 amino acid motif, the F box. The F box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1 cullin F box), which function in phosphorylation dependent ubiquitination. The F box proteins are divided into 3 classes: Fbws containing WD 40 domains, Fbls containing leucine rich repeats, and Fbxs containing either different protein protein interaction modules or no recognizable motifs. FBXO25 belongs to the Fbxs class. There are three named isoforms produced by alternative splicing.FBXO25, also known as FBX25, is a 367 amino acid protein that contains one C-terminal F-box domain and belongs to the Fbx class of the F-box family of proteins. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and recruitment for ubiquitination. They are members of a larger family of proteins that are involved in the regulation of a wide variety of cellular processes (including the cell cycle, immune response, signaling cascades and developmental processes) through the targeting of proteins, such as cyclins, cyclin-dependent kinase inhibitors, IkB-a and b-catenin, for degradation by the proteasome after ubiquitination. Expressed at high levels in brain, FBXO25 localizes predominantly to the nucleus and directly interacts with Skp1 p19 and CUL-1. Disruption of the gene encoding FBXO25 can lead to X-linked mental retardation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXO25
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR