Anti-CLDN7 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-8482R-CY5
10485-564EA 792.04 USD
10485-564
Anti-CLDN7 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Plays a major role in tight junction-specific obliteration of the intercellular space.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLDN7
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR