Anti-HLA-DMB Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-8481R-A488
10485-530EA 792.04 USD
10485-530
Anti-HLA-DMB Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Plays a critical role in catalyzing the release of class II-associated invariant chain peptide (CLIP) from newly synthesized MHC class II molecules and freeing the peptide binding site for acquisition of antigenic peptides. In B-cells, the interaction between HLA-DM and MHC class II molecules is regulated by HLA-DO.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HLA-DMB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR