Anti-FBXL17 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-8469R-FITC
10485-298EA 792.04 USD
10485-298
Anti-FBXL17 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
FBXL17

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXL17
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR