Anti-FBXL3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-8467R-A488
10485-234EA 792.07 USD
10485-234
Anti-FBXL3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Substrate-recognition component of some SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex involved in circadian clock function. The SCF(FBXL3) complex acts by mediating ubiquitination and subsequent degradation of CRY1 and CRY2. Recruiter of target protein that may recognize and bind to some phosphorylated proteins and promotes their ubiquitination and degradation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR