Anti-APLP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-8465R-A647
10485-148EA 792.04 USD
10485-148
Anti-APLP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
May play a role in postsynaptic function. The C-terminal gamma-secretase processed fragment, ALID1, activates transcription activation through APBB1 (Fe65) binding (By similarity). Couples to JIP signal transduction through C-terminal binding. May interact with cellular G-protein signaling pathways. Can regulate neurite outgrowth through binding to components of the extracellular matrix such as heparin and collagen I.The gamma-CTF peptide, C30, is a potent enhancer of neuronal apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APLP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR