Anti-SENP5 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-8464R-CY5.5
10484-788EA 792.04 USD
10484-788
Anti-SENP5 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO3 to its mature form and deconjugation of SUMO2 and SUMO3 from targeted proteins. Has weak proteolytic activity against full-length SUMO1 or SUMO1 conjugates. Required for cell division.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SENP5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR