Anti-APLP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-8462R-FITC
10484-760EA 792.04 USD
10484-760
Anti-APLP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
APLP2 is a human sperm membrane protein which contains a segment with high homology to the transmembrane-cytoplasmic domains of APP found in brain plaques of Alzheimer disease patients. The human amyloid precursor-like protein APLP2 is a highly conserved homolog of a sequence-specific DNA-binding mouse protein with an important function in the cell cycle. The gene which encodes APLP2 maps to human chromosome 11q24.May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3'(CDEI box). Inhibits trypsin, chymotrypsin, plasmin, factor XIA and plasma and glandular kallikrein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APLP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR