Anti-DCUN1D4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-8251R-A488
10480-256EA 792.04 USD
10480-256
Anti-DCUN1D4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The DCN1-like protein family is comprised of Dcun1D1, Dcun1D2, Dcun1D3, Dcun1D4 and Dcun1D5. The founding member, Dcun1D1, is involved in the malignant transformation of squamous cell lineage.Dcun1D4, (defective in cullin neddylation protein 1-like protein 4 or DCN1-like protein 4), also designated KIAA0276, exists as 2 isoforms as a result of alternative splicing and contains one DCUN1 domain. The gene encoding Dcun1D4 maps to chromosome 4, which houses nearly 6% of the human genome and has the largest gene deserts (regions of the genome with no protein encoding genes) of all of the human chromosomes. Defects in some of the genes located on chromosome 4 are associated with Huntington's disease, Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DCUN1D4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR