Anti-DCAKD Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-8249R-CY7
10480-222EA 792.04 USD
10480-222
Anti-DCAKD Rabbit Polyclonal Antibody (Cy7®)
Antibodies
DCAKD belongs to the coaE family. It contains one DPCK (dephospho CoA kinase) domain. There are two isoforms.Coenzyme A (CoA) is an essential cofactor used in numerous biochemical pathways. It plays a critical role in the synthesis and oxidation of fatty acids and is vital to the citric acid cycle. The biosynthesis pathway of CoA from pantothenic acid (also known as vitamin B5) is essential and universal in prokaryotes and eukaryotes. In humans, the final steps of the biosynthesis pathway are carried out by the bifunctional enzyme COASY. The sequence of these enzymes are highly conserved between different bacterial species. The phosphopantetheine adenylyltransferase and decoenzyme A kinase activities of COASY are evolutionarily conserved activities. DCAKD (deCoA kinase domain containing protein) is a 231 amino acid protein that consists of a deCoA kinase domain and an ATP nucleotide binding motif. Localizing to mitochondria and the cytosol, DCAKD belongs to the coaE family which suggests that it may play a role in the biosynthesis of CoA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DCAKD
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR