Anti-CUTA Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-8248R-A350
10479-950EA 792.07 USD
10479-950
Anti-CUTA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
May forms part of a complex of membrane proteins attached to acetylcholinesterase (AChE).Tissue specificity:Ubiquitous. Widely expressed in brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CUTA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR