Anti-CADM2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-8246R-FITC
10479-726EA 792.04 USD
10479-726
Anti-CADM2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Adhesion molecule that engages in homo- and heterophilic interactions with the other nectin-like family members, leading to cell aggregation. Important for synapse organization, providing regulated trans-synaptic adhesion. Preferentially binds to oligodendrocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CADM2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR