Anti-CSN1S1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-8245R-HRP
10480-184EA 792.05 USD
10480-184
Anti-CSN1S1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Important role in the capacity of milk to transport calcium phosphate.Casoxin D acts as opioid antagonist and has vasorelaxing activity mediated by bradykinin B1.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CSN1S1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR