Anti-CSN1S1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-8245R-CY5.5
10480-178EA 792.05 USD
10480-178
Anti-CSN1S1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Important role in the capacity of milk to transport calcium phosphate.Casoxin D acts as opioid antagonist and has vasorelaxing activity mediated by bradykinin B1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSN1S1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR