Anti-CA11 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-8244R-CY7
10480-124EA 792.04 USD
10480-124
Anti-CA11 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide. CAs show extensive diversity in their distribution and subcellular localization. They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA XI, also referred to as carbonic anhydrase-related protein 11 precursor (CA-RP XI) or carbonic anhydrase-related protein 2 (CA-RP II), is a member of the carbonic anhydrase family that lacks two of the three Zn-binding motifs essential for carbonic anhydrase activity. For this reason, CA XI does not exhibit catalytic activity. It is expressed primarily in brain but is also found in spinal cord and thyroid. CA XI may play a role in brain development.Tissue specificity:Expressed abundantly in the brain with moderate expression also present in spinal cord and thyroid.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA11
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR