Anti-FAM164B Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-8227R-CY5.5
10479-698EA 792.04 USD
10479-698
Anti-FAM164B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
No data available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM164B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR