Anti-FAM164B Rabbit Polyclonal Antibody (Biotin)

BS-8227R-BIO
10479-692EA 529.29 USD
10479-692
Anti-FAM164B Rabbit Polyclonal Antibody (Biotin)
No data available.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: FAM164B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR