Anti-FAM164B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-8227R-A488
10479-686EA 792.04 USD
10479-686
Anti-FAM164B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
No data available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM164B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR