Anti-GAREML Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-8217R-FITC
10479-486EA 792.04 USD
10479-486
Anti-GAREML Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
FAM59B

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM59B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR