Anti-KLHL25 Rabbit Polyclonal Antibody (Cy5.5®)

BS-8051R-CY5.5
10475-724EA 592.02 USD
10475-724
Anti-KLHL25 Rabbit Polyclonal Antibody (Cy5.5®)
KLHL25

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHL25
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR