Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NCAPG2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-7721R-A647
10468-194
Anti-NCAPG2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
NCAPG2 is 1 of 3 non-SMC subunits that define condensin II. Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 and SMC4, but they contain different sets of non-SMC subunits. NCAPG2 is 1 of 3 non-SMC subunits that defines condensin II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCAPG2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty