Anti-NCAPG2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7721R-A350
10468-188EA 792.04 USD
10468-188
Anti-NCAPG2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
NCAPG2 is 1 of 3 non-SMC subunits that define condensin II. Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 and SMC4, but they contain different sets of non-SMC subunits. NCAPG2 is 1 of 3 non-SMC subunits that defines condensin II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCAPG2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR