Anti-DSCC1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7720R-CY3
10468-174EA 792.04 USD
10468-174
Anti-DSCC1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Loads PCNA onto primed templates regulating velocity, spacing and restart activity of replication forks. May couple DNA replication to sister chromatid cohesion through regulation of the acetylation of the cohesin subunit SMC3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DSCC1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR