Anti-CDCA5 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7717R-CY3
10468-086EA 792.04 USD
10468-086
Anti-CDCA5 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Regulator of sister chromatid cohesion in mitosis stabilizing cohesin complex association with chromatin. May antagonize the action of WAPAL which stimulates cohesin dissociation from chromatin. Cohesion ensures that chromosome partitioning is accurate in both meiotic and mitotic cells and plays an important role in DNA repair. Required for efficient DNA double-stranded break repair.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDCA5
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR