Anti-NUF2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7714R-CY3
10468-014EA 792.05 USD
10468-014
Anti-NUF2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Acts as a component of the essential kinetochore-associated NDC80 complex, which is required for chromosome segregation and spindle checkpoint activity. Required for kinetochore integrity and the organization of stable microtubule binding sites in the outer plate of the kinetochore.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NUF2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR