Anti-RAI17 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-7709R-CY7
10467-900EA 792.04 USD
10467-900
Anti-RAI17 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Increases ligand-dependent transcriptional activity of AR and promotes AR sumoylation. The stimulation of AR activity is dependent upon sumoylation.Tissue specificity:Expressed most abundantly in ovary and, at lower levels, in prostate, spleen and testis. Weak expression, if any, in thymus, small intestine, colon and peripheral blood leukocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAI17
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR