Anti-ZNF415 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7708R-A350
10467-652EA 792.04 USD
10467-652
Anti-ZNF415 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Involved in transcriptional regulation. Transcriptional activity differed among the various isoforms. All isoforms except isoform 3 seem to suppresses the transcriptional activities of AP-1 and p53/TP53.Tissue specificity:Expressed in all tissues examined. Isoforms are differentially expressed. Isoform 3 and isoform 5 were highly expressed, isoform 4 moderately expressed, isoform 2 lower expression, the lowest expression level was seem with isoform 1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF415
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR