Anti-TSPYL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-7703R-HRP
10467-864EA 792.04 USD
10467-864
Anti-TSPYL5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Involved in modulation of cell growth and cellular response to gamma radiation probably via regulation of the Akt signaling pathway. Involved in regulation of p53/TP53. Suppresses p53/TP53 protein levels and promotes its ubiquitination; the function is dependent on USP7 and independent on MDM2. Proposed to displace p53/TP53 from interaction with USP7.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TSPYL5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR