Anti-SLC12A3 Rabbit Polyclonal Antibody (Biotin)

BS-7694R-BIO
10467-618EA 545.21 USD
10467-618
Anti-SLC12A3 Rabbit Polyclonal Antibody (Biotin)
Key mediator of sodium and chloride reabsorption in this nephron segment, accounting for a significant fraction of renal sodium reabsorption.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SLC12A3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR