Anti-Relaxin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-7692R-A555
10467-566EA 792.04 USD
10467-566
Anti-Relaxin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Relaxin is a peptide hormone produced by the corpora lutea of ovaries during pregnancy in many mammalian species, including man. The secretion of the hormone into the blood stream just before parturition results in a marked softening and lengthening of the pubic symphysis and a softening of the cervix, which facilitates the birth process. By inhibiting uterine contractions, relaxin may influence the timing of parturition. Like insulin, relaxin consists of two peptide chains, A and B, covalently linked by disulfide bonds. By further analogy to insulin, the two peptides are synthesized as a single-chain precursor polypeptide with the B chain at the NH2-terminus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Relaxin 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR