Anti-ATP6AP2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-7691R-CY7
10467-554EA 746.17 USD
10467-554
Anti-ATP6AP2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Functions as a renin and prorenin cellular receptor. May mediate renin-dependent cellular responses by activating ERK1 and ERK2. By increasing the catalytic efficiency of renin in AGT/angiotensinogen conversion to angiotensin I, it may also play a role in the renin-angiotensin system (RAS).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP6AP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR