Anti-KCNMB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-7689R-A647
10467-520EA 792.04 USD
10467-520
Anti-KCNMB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open. Maxi K+ beta is the target for possible therapeutics because of its role in blood flow and blood pressure regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KCNMB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR