Anti-CORIN Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7685R-A350
10467-246EA 792.05 USD
10467-246
Anti-CORIN Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Serine-type endopeptidase involved in atrial natriuretic peptide hormone (NPPA) processing. Converts through proteolytic cleavage the non-functional propeptide NPPA into the active hormone, thereby regulating blood pressure in heart and promoting natriuresis, diuresis and vasodilation. Proteolytic cleavage of pro-NPPA also plays a role in female pregnancy by promoting trophoblast invasion and spiral artery remodeling in uterus. Also acts as a regulator of sodium reabsorption in kidney. May also process pro-NPPB the B-type natriuretic peptide. Isoform 2: has weaker endopeptidase activity compared to isoform 1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CORIN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR