Anti-TReP132 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-7683R-A555
10467-378EA 792.04 USD
10467-378
Anti-TReP132 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Activates transcription of CYP11A1. Interaction with CREBBP and EP300 results in a synergistic transcriptional activation of CYP11A1.Tissue specificity:Highest expression was seen in thymus, testis and adrenal cortex, expressed also in the adrenal medulla, thyroid, and stomach. Highly expressed in steroidogenic JEG-3 and MCF-7 cells, low expression was seen in non-steroidogenic HepG2 and HK293 cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TReP132
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR