Anti-FKBPL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-7681R-HRP
10467-202EA 792.04 USD
10467-202
Anti-FKBPL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May be involved in response to X-ray. Regulates p21 protein stability by binding to Hsp90 and p21.Tissue specificity: Ubiquitously expressed with higher levels in testis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: WISP39
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR