Anti-BCCIP Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-7679R-CY5.5
10467-170EA 792.04 USD
10467-170
Anti-BCCIP Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
FunctionMay promote cell cycle arrest by enhancing the inhibition of CDK2 activity by CDKN1A. May be required for repair of DNA damage by homologous recombination in conjunction with BRCA2. May not be involved in non-homologous end joining.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCCIP
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR