Anti-ZDHHC16 Rabbit Polyclonal Antibody (Biotin)

BS-7677R-BIO
10467-068EA 529.29 USD
10467-068
Anti-ZDHHC16 Rabbit Polyclonal Antibody (Biotin)
Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: ZDHHC16
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR