Anti-ZDHHC16 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-7677R-A555
10467-064EA 792.04 USD
10467-064
Anti-ZDHHC16 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZDHHC16
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR