Anti-TMEM123 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-7674R-A555
10467-014EA 792.04 USD
10467-014
Anti-TMEM123 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
This gene encodes a highly glycosylated transmembrane protein with a high content of threonine and serine residues in its extracellular domain, similar to a broadly defined category of proteins termed mucins. Exposure of some cell types to anti PORIMIN (pro oncosis receptor inducing membrane injury) antibody, crosslinks this protein on the cell surface and induces a type of cell death termed oncosis. Oncosis is distinct from apoptosis and is characterized by a loss of cell membrane integrity without DNA fragmentation. This gene product is proposed to function as a cell surface receptor that mediates cell death.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMEM123
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR