Anti-GNRH2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-7672R-A488
10466-988EA 792.04 USD
10466-988
Anti-GNRH2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
A single transmembrane protein highly expressed in embryonic tissues and involved in inducing apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NRH2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR