Anti-GPR65 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-7668R-CY3
10466-922EA 658.44 USD
10466-922
Anti-GPR65 Rabbit Polyclonal Antibody (Cy3®)
Receptor for the glycosphingolipid psychosine (PSY) and several related glycosphingolipids. May have a role in activation-induced cell death or differentiation of T-cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR65
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR