Anti-SLTM Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-7667R-A350
10466-888EA 792.04 USD
10466-888
Anti-SLTM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
When SLTM is overexpressed it acts as a general inhibitor of transcription that eventually leads to apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLTM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR